Imprimer
CO₂ incubator shaker, S41i
Fournisseur: New Brunswick
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
444-1092EA 0 EUR
390-0811 444-1092
CO₂ incubator shaker, S41i
Agitateurs Agitateurs incubateurs
The S41i CO₂ incubator is specifically designed for non-adherent cells. It combines precise temperature and CO₂ control with an excellent laboratory shaker to give high cell yields and viability. The six-sided direct heating profile provides exceptionally uniform temperature and incubator environment; it guards against wide fluctuations in temperature and CO₂ that can shock cells. The sealed inner and outer doors and advanced PI control maintain temperature accuracy and uniformity while minimising gas consumption.
  • Gentle, fanless convection circulation allows full use of the incubator interior
  • Easy to read LED display for temperature, CO₂ and shaking speed
  • IR CO₂ sensor with automatic auto-zero to ensure accurate calibrated measurements
  • Stackable – two units high with optional stacking kits
  • Four eccentric counterbalanced drives and brushless motor ensure stable, uniform motion
  • Integral timer, 0,1 to 99,9 h with automatic switch-off and audible alarm when programme is complete
Units feature a LED display for temperature, CO₂ and shaking speed. The chamber is constructed from seamless stainless steel for easy cleaning, with one removable shelf. The sealed inner glass door allows sample viewing, whilst maintaining environmental integrity. The S41i comes with a 25 mm access port and USB communications port. A high-temperature disinfection cycle protects against bacterial contamination. HEPA filtration of gas supply inlets ensures sterility. Audio/visual alarms for temperature, CO₂ and door open.
Informations sur les accessoires: Note: S41i accommodates a maximum of 20×125, 16×250, 12×500, 10×1000, 4×2000, 3×2800 or 3×4000 ml flasks using the universal platform, and 32×125, 25×250, 15×500, 11×1000, 6×2000, 3×2800 or 3×4000 ml flasks using dedicated platforms.
Informations de livraison: Supplied with one perforated stainless steel shelf, but without platforms or accessories, required accessories must be ordered separately. For details of accessories, including shaking platforms, additional shelves and stacking kits, please contact your local Avantor office. BioCommand® SFI software is available for remote and historical data logging and report generation.
Attention: * with 25 mm orbit