Imprimer
Single element standards for ICP-OES/MP-AES
Fournisseur: Agilent Technologies
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
VARI190026432EA 0 EUR
VARI190026432 VARI190026414 VARI190026419 VARI190026429 VARI190026441 VARI190026435 VARI190026430 VARI190026425 VARI190026437 VARI190026436 VARI190026421 VARI190026431 VARI190026417 VARI190026415 VARI190026418 VARI190026426 VARI190026420 VARI190026433 VARI190026439 VARI190026424 VARI190026423 VARI190026440 VARI190026449
Single element standards for ICP-OES/MP-AES
Solutions are manufactured using high purity starting materials, which are regularly tested in accordance with NIST guidelines to provide traceability.
  • Packaged in a pre-leached HDPE bottle, secured with a tamper-evident seal
  • Certificates of analysis supplied