Imprimer
Bottles, narrow neck, square, HDPE, with screw cap
Fournisseur: Vitlab
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
VITL128693EA 0 EUR
VITL128693 VITL128793 VITL128493 VITL128593
Bottles, narrow neck, square, HDPE, with screw cap
Flacons Flacons en plastique
HDPE, translucent.
  • Space saving
  • Good chemical resistance
  • Good temperature resistance
With PP screw cap.