Imprimer
Plaques HPTLC, nano gel de silice 60, couches non modifiées, Nano-SIL
Fournisseur: MACHEREY-NAGEL
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
ALUGRAM®
552-0330EA 341 EUR
552-0330 552-0230 MANA818240 MANA818241 552-0301 552-0228 552-0227 552-0303 552-0226 552-0225 MANA818342 552-0229
Plaques HPTLC, nano gel de silice 60, couches non modifiées, Nano-SIL
Plaques CCM
Unmodified HPTLC silica layers for HPTLC.
  • Nano silica gel (particle size 2 to 10 μm)
  • Narrow fractionation of the silica particles
  • Binders are stable in almost all organic solvents
Nano-SIL HPTLC plates and sheets allow sharper separations, shorter developing times and an increased detection sensitivity compared to standard silica gel plates.
Nano silica 60: Specific surface (BET) ~ 500 m²/g; specific pore volume 0,75 ml/g; particle size 2 to 10 μm.