Imprimer

3-Chloro-4-fluoro-N-(3-fluorobenzyl)aniline, 97%

Ratings: Ecrire un avis

Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5

H57733.03EA 391 EUR
ALFAH57733.03 ALFAH57733.MD
3-Chloro-4-fluoro-N-(3-fluorobenzyl)aniline, 97%
Purity: 97%
CAS No.: 1040309-97-4
Molecular Formula: C13H10ClF2N
Formula Weight: 253.67
MDL No.: MFCD11180481
Commander maintenant

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR