Imprimer

4-Chloro-2-fluoro-N-(4-fluorobenzyl)aniline, 97%

Ratings: Ecrire un avis

Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5

H57994.MDEA 69.8 EUR
ALFAH57994.MD ALFAH57994.03
4-Chloro-2-fluoro-N-(4-fluorobenzyl)aniline, 97%
Purity: 97%
CAS No.: 1152635-08-9
Molecular Formula: C13H10ClF2N
Formula Weight: 253.67
Commander maintenant

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR