Imprimer

N-(4-Fluorobenzyl)-4-iodoaniline, 97%

Ratings: Ecrire un avis

Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5

H57036.MDEA 88.7 EUR
ALFAH57036.MD ALFAH57036.03
N-(4-Fluorobenzyl)-4-iodoaniline, 97%
Purity: 97%
CAS No.: 1040018-16-3
Molecular Formula: C13H11FIN
Formula Weight: 327.14
MDL No.: MFCD11179992
Commander maintenant

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR