Imprimer
HPTLC plates, nano silica gel 60, modified layers, Nano-SIL NH₂
Fournisseur: MACHEREY-NAGEL
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
ALUGRAM®
552-0255EA 708 EUR
552-0255 552-0322 552-0254
HPTLC plates, nano silica gel 60, modified layers, Nano-SIL NH₂
Plaques CCM
Amino-modified HPTLC silica layers are available as glass plates or ALUGRAM® aluminium sheets. They are ideal for the separation of vitamins, sugars, steroids, purine derivatives, xanthines, phenols, nucleotides, and pesticides.
  • Layer can be wetted equally well by pure water as by organic solvents
Base material: Nano silica 60: Specific surface (BET) ~ 500 m²/g; specific pore volume 0,75 ml/g; particle size 2 to 10 μm; pH stability 2 to 8.
Modification: Aminopropyl, carbon content 3,5%
Indicator: Acid-resistant product with a pale blue fluorescence for short-wave UV (254 nm), and UV-absorbing substances appear as dark-blue to black spots on a light-blue background.