Imprimer
Capillary micropipettes, Acura® 841
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
SOCO841.005EA 324 EUR
SOCO841.005 613-0550
Capillary micropipettes, Acura® 841
Pipette with ETFE tipped plunger for positive displacement of the sample in a precision capillary tube. Optimal accuracy and reproducibility when pipeting high density, viscous, foaming or volatile liquids.
  • Step-wise volume adjustment
  • Interchangeable plunger and glass capillary
  • Matching colour codes on pipette, plunger and capillary
  • Stainless steel, ETFE tipped plunger