Imprimer
Safety shoes, lace-up, Jalosiris
Fournisseur: Jallatte
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
Jalosiris
JALCJ0646/47EA 0 EUR
JALCJ0646/47 JALCJ0646/48 JALCJ0646/44 JALCJ0646/42 JALCJ0646/45 JALCJ0646/40 JALCJ0646/46 JALCJ0646/43 JALCJ0646/39 JALCJ0646/41
Safety shoes, lace-up, Jalosiris
Chaussures et bottes
Metal-free safety shoes made from full water repellent grain leather upper, 3D lining and Softane™/Néotril™ sole (heat resistant up to 300 °C); with padded tongue.
  • Lining with three dimensional foam structures for excellent ventilation
  • Anatomically shaped Confort-Tech insole; anti-static and antibacterial, with shock absorber heel
  • Slip resistant Triftane™ sole helps to prevent fatigue
  • Lightweight, non magnetic, perforated composite toecap (200 J)
  • Non magnetic, lightweight and flexible FleXtane™ HP midsole (100% composite)
Colour: Black
Certifications: EN ISO 20345:2004; S3, HRO, SRC