Imprimer
ddNTP, 100 mM solutions
Fournisseur: Cytiva
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
27-2081-01EA 315 EUR
27-2081-01 27-2061-01
ddNTP, 100 mM solutions
Réactifs pour acides nucléiques Nucléotides
Easy to use, time saving solutions for use in any application requiring ddNTPs of high purity, such as dideoxy sequencing and genotyping reactions.
  • High purity ≥98% triphosphate
  • Each lot of ddNTPs is functionally tested to meet or exceed criteria for high quality sequencing