Imprimer
Colonnes capillaires, PERMABOND® SE-54 HKW
Fournisseur: MACHEREY-NAGEL
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
Permabond®
MANA723945.25EA 870 EUR
MANA723945.25 MANA723945.50
Colonnes capillaires, PERMABOND® SE-54 HKW
Colonnes pour chromatographie
Special GC phase for the analysis of volatile halogenated hydrocarbons.
  • Optimised nonpolar phase for volatile halogenated hydrocarbons
  • Max. temperature for isothermal operation 300 °C
  • Max. temperature for short isotherms in a temperature program 320 °C
USP code: G36.