Imprimer

2-Chloro-4-(3,5-dimethylphenylcarbamoyl)benzeneboronic acid, 97%

Ratings: Ecrire un avis

Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5

H54549.03EA 326 EUR
ALFAH54549.03 ALFAH54549.MD
2-Chloro-4-(3,5-dimethylphenylcarbamoyl)benzeneboronic acid, 97%
Purity: 97%
Molecular Formula: C15H15BClNO3
Formula Weight: 303.55
MDL No.: MFCD22125075
Commander maintenant

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR