Imprimer

2-Chloro-4-(2-methyl-4-nitrophenylcarbamoyl)benzeneboronic acid, 97%

Ratings: Ecrire un avis

Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5

H55202.MDEA 123 EUR
ALFAH55202.MD ALFAH55202.03
2-Chloro-4-(2-methyl-4-nitrophenylcarbamoyl)benzeneboronic acid, 97%
Purity: 97%
Molecular Formula: C14H12BClN2O5
Formula Weight: 334.52
MDL No.: MFCD22125124
Commander maintenant

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR