Imprimer

3-[Benzyl(methyl)carbamoyl]-5-fluorobenzeneboronic acid, 97%

Ratings: Ecrire un avis

Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5

H54520.03EA 391 EUR
ALFAH54520.03 ALFAH54520.MD
3-[Benzyl(methyl)carbamoyl]-5-fluorobenzeneboronic acid, 97%
Purity: 97%
Molecular Formula: C15H15BFNO3
Formula Weight: 287.10
MDL No.: MFCD20040213
Commander maintenant

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR