Imprimer

5-[Benzyl(ethyl)carbamoyl]-2-fluorobenzeneboronic acid, 97%

Ratings: Ecrire un avis

Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5

H54357.03EA 391 EUR
ALFAH54357.03 ALFAH54357.MD
5-[Benzyl(ethyl)carbamoyl]-2-fluorobenzeneboronic acid, 97%
Purity: 97%
Molecular Formula: C16H17BFNO3
Formula Weight: 301.12
MDL No.: MFCD20040173
Commander maintenant

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR