Imprimer

3-Fluoro-5-(4-methoxybenzylcarbamoyl)benzeneboronic acid, 97%

Ratings: Ecrire un avis

Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5

H54829.MDEA 97.6 EUR
ALFAH54829.MD ALFAH54829.03
3-Fluoro-5-(4-methoxybenzylcarbamoyl)benzeneboronic acid, 97%
Purity: 97%
Molecular Formula: C15H15BFNO4
Formula Weight: 303.10
MDL No.: MFCD20265351
Commander maintenant

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR