Imprimer
Glycerol, Molecular Biology Grade
Fournisseur: Thermo Scientific
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
J61059.K2EA 161 EUR
ALFAJ61059.K2 ALFAJ61059.AE ALFAJ61059.AP
Glycerol, Molecular Biology Grade
CAS No.: 56-81-5
Molecular Formula: C3H8O3
Formula Weight: 92.09
Flash Point: 160°(320°F)
Physical Form: Liquid
Density: 1.26
Refractive Index: 1.474
Sensitivity: Hygroscopic
Appearance: Colourless
Odour: Odourless
Melting Point: 18°
Boiling Point: 182°/20mm
Solubility: Miscible with water
Beilstein Reference: 635685
Merck Reference: 14,4484
MDL No.: MFCD00004722
Useful as a cryoprotectant