Imprimer
Roll top hardcover spill pallet, PIG®
Fournisseur: New Pig
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
PIG®
NEWPPAK601-BWG-AWOEA 1230 EUR
NEWPPAK601-BWG-AWO NEWPPAK901-BWG-AWO NEWPPAK601-AWO
Roll top hardcover spill pallet, PIG®
Stockage des fûts
These roll top hardcovers can be used for all-weather protection and liquid containment. Durable swing-out doors on front offers easy loading and unloading.
  • Weather-resistant design prevents contaminants from entering stormwater
  • Heavy-duty construction allows hardcover and pallet to be lifted by a forklift with drums in place
  • Low-density polyethylene (LDPE) with UV inhibitors resists UV rays, rust, corrosion and most chemicals for long, trouble-free life
  • Doors and roll top are available with a padlock