Imprimer
Fluorographic reagent, Amersham Amplify™
Fournisseur: Cytiva
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
NAMP100EA 404 EUR
GEHENAMP100
Fluorographic reagent, Amersham Amplify™
Amplify™ is a fast and convenient fluorographic reagent for ³⁵S-, ¹⁴C- and ³H-labelled samples in gels. Amplify™ can increase sensitivity of detection and significantly reduce exposure times required for weak beta emitters.
  • Increase detection efficiency 1000-fold for ³H and 15-fold for ³⁵S and ¹⁴C
  • Less than 2 hours of total preparation time
  • Can be used with acrylamide or agarose gels with no change in protocols
  • Odourless and inorganic