Imprimer
Mammalian Protein Extraction Buffer
Fournisseur: Cytiva
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
28-9412-79EA 75.4 EUR
GEHE28-9412-79
Mammalian Protein Extraction Buffer
Mammalian protein extraction buffer is a 500 ml solution designed for efficient and gentle extraction of biologically active, total soluble proteins from mammalian cultured cells. This buffer is based on organic buffering agents, mild non ionic detergents, and a combination of various salts and agents to gently disrupt the cell wall and release soluble proteins.
Mammalian protein extraction buffer is compatible with most applications, including enzyme assays, various chromatography procedures, and electrophoresis. After preparation, the sample may be used for most enzyme assays including reporter gene assays, kinases, and immunoassays.