Imprimer
Ion exchange chromatography media, CM Sephadex™
Fournisseur: Cytiva
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
Sephadex™
17-0220-02EA 1920 EUR
17-0220-02 17-0210-02 17-0210-01 17-0220-01
Ion exchange chromatography media, CM Sephadex™
Protéines purification Protein Purification Resins
CM Sephadex™ is a weak cation exchanger based on the well documented and well proven Sephadex™ base matrix.
  • Weak cation exchanger suitable batch techniques
  • High binding capacities