Imprimer
Affinity purification media, GammaBind™ G Sepharose™
Fournisseur: Cytiva
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
Sepharose™
17-0885-01EA 835 EUR
17-0885-01 17-0885-02
Affinity purification media, GammaBind™ G Sepharose™
Milieux de purification des protéines
GammaBind™ G Sepharose™ is GammaBind™ G, type 2, covalently immobilised to Sepharose™ 4B by the cyanogen bromide method. It is an affinity medium for laboratory scale antibody purification.
  • Binds to all IgG subclasses from human, mouse, and rat
  • Binds total IgG from guinea pig, rabbit, goat, cow sheep, and horse
  • Will not cross-react with other serum proteins such as IgM, IgE, IgA or transferrin
It binds to the Fc region of IgG from a variety of mammalian species. It may be used to analyse and purify classes, subclasses and fragments of immunoglobulins from any biological fluid or cell culture medium. Extremely useful for isolation of immune complexes.