Imprimer
Affinity chromatography media, 2'5' ADP Sepharose™ 4B
Fournisseur: Cytiva
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
Sepharose™
17-0700-01EA 1020 EUR
GEHE17-0700-01
Affinity chromatography media, 2'5' ADP Sepharose™ 4B
2'5' ADP Sepharose™ 4B media with immobilised NADP structural analogue is primarily used for purification of enzymes requiring NADP as a cofactor. Selective elution with gradients of NAD⁺ or NADP⁺ has allowed the resolution of complex mixtures of dehydrogenase isoenzymes using 2'5' ADP Sepharose™ 4B.