Imprimer
Reversed phase FPLC columns, SOURCE™ 5RPC ST
Fournisseur: Cytiva
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
17-5116-01EA 1040 EUR
GEHE17-5116-01
Reversed phase FPLC columns, SOURCE™ 5RPC ST
SOURCE™ 5RPC ST column designed for analytical and preparative reversed phase chromatography (RPC). Suitable for LC-MS applications. It is prepacked with SOURCE™ 15RPC.
  • For high-resolution analytical separations as required for peptide mapping and LC-MS techniques
  • Superior resolution in the pH range 1 - 12
  • Long-life column due to high chemical and physical resistance
  • High column-to-column reproducibility