Imprimer
Système de nettoyage, Hygen™
Fournisseur: Rubbermaid Commercial Products
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
FGQ966000000 FGQ96958YL00 R050669 1861160
141-0136EA 127 EUR
141-0136 141-0135 141-0140 RUBB1861160
Système de nettoyage, Hygen™
Serpillières
HYGEN™ PULSE™ spray mop provides an ergonomic, bucket-less mopping system to help cover more square metres in less time than traditional systems.
  • Patented onboard reservoir and solution system evenly dispenses three sprays of cleaner behind mop
  • Unscrew the cap to manually refill or connect to the PULSE™ Caddy hose for automatic refills
  • Holds up to 0,6 L of liguid solution to clean up to 80 m² of floor space
  • Non-slip tip helps prevent the hanlde from sliding and falling when placed against a wall
The HYGEN™ PULSE™ high-capacity caddy allows users to easily clean more square meters, reducing the frequency of trips to the supply closet. The 7,5 L capacity will refill the Rubbermaid PULSE™ spray mop up to 12 times. The caddy is designed to fit in Rubbermaid cleaning carts for efficient on-the-go refills.