Imprimer
Reversed phase HPLC columns, µRPC C2/C18
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
17-5057-01EA 0 EUR
GEHE17-5057-01
Reversed phase HPLC columns, µRPC C2/C18
µRPC C2/C18 is a prepacked stainless steel column for fast, sensitive, and highest-resolution reversed phase chromatography at microscale. Porous silica beads with a mixture of C2 and C18 alkyl ligands.
  • The small bead size for exceptional resolution
  • Excellent choice for very high resolution analysis and preparative micropurification
  • High-efficiency medium for peptide mapping and analysis requiring very high resolution
  • Available in a prepacked column format for ÄKTAdesign and other high-performance liquid chromatography systems