Imprimer
N,N'-Methylene-bisacrylamide, PlusOne™
Fournisseur: Cytiva
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
17-1304-02EA 174 EUR
GEHE17-1304-02
N,N'-Methylene-bisacrylamide, PlusOne™
PlusOne™ N,N'-Methylenebisacrylamide is a general cross-linker used with acrylamide for PAGE techniques. It is supplied as a dry powder.