Imprimer
SWIFT II Application software, for 21 CFR part 11 complaints
Fournisseur: Cytiva
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
80-2117-57EA 0 EUR
GEHE80-2117-57
SWIFT II Application software, for 21 CFR part 11 complaints
SWIFT II Applications Software is 21 CFR part 11 compliant in terms of electronic records and signatures with the flexibility and functionality of the established SWIFT II software, suitable for any analytical or QC laboratory that operates within such an environment. The software comprises both client and server applications for networked installations, but both of these can be installed on one PC if required.
  • Full password protection; electronic records and signatures compliant
  • Full audit trail
  • All the functionality of standard SWIFT II Method software
  • Compatible with Ultrospec 2100, 3100, 3300, 4300, 5300 and 6300 pro
When combined with the fully Pharmacopoeia compliant Ultrospec 6300 pro and 5300 pro instruments, extremely powerful and sophisticated systems are generated. Note that as in any 21 CFR part 11 compliant environment, it is incumbent on the end user to have the necessary standard operating procedures (SOPs) and training in place to ensure optimal use of the equipment.