Imprimer
Cytokine and growth factor assays, Biotrak™ ELISA
Fournisseur: Cytiva
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
RPN2779EA 1040 EUR
GEHERPN2779 GEHERPN2742 GEHERPN2764
Cytokine and growth factor assays, Biotrak™ ELISA
The Biotrak™ cytokine and growth factor ELISAs allow the convenient measurement of human, rat, and mouse cytokine protein levels in serum, plasma, and cell culture samples. High-sensitivity human cytokine ELISAs that utilise Amdex multi-enzyme conjugates for signal amplification are also available.
  • Assays specific for human, rat or mouse
  • Colorimetric readout in 96-well microplate format
  • Suitable for use in a range of sample types