Imprimer
Protein kinase C Biotrak™ enzyme assay
Fournisseur: Cytiva
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
RPN77EA 737 EUR
GEHERPN77
Protein kinase C Biotrak™ enzyme assay
Protein Kinase C Enzyme Assay has been developed to allow researchers to quantitatively measure PKC activity in a wide range of samples, in a convenient and reproducible manner.
  • Enzyme immunoassay
  • Suitable for use with tissue and cell preparations
  • To be used with [g–32P]ATP or [g–33P]ATP
Informations de livraison: Includes calcium buffer, lipid, peptide buffer, DTT buffer, magnesium ATP buffer, stop reagent and peptide binding papers.