Imprimer
Blood collecting tubes with serum gel (clotting activator), S-Monovette®
Fournisseur: Sarstedt
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
S-Monovette®
SARS01.1602.001EA 418 EUR
SARS01.1602.001 SARS04.1905.001-50 SARS06.1667.001-50 SARS01.1602.001_50 SARS03.1524 SARS01.1602.00 SARS04.1905.001 SARS04.1935.001 SARS02.1388 SARS04.1935.001-50 SARS02.1388-50 SARS04.1905 SARS02.1388.001 SARS04.1935 SARS03.1524.001 SARS06.1667.001
Blood collecting tubes with serum gel (clotting activator), S-Monovette®
Systèmes de diagnostic clinique Prélèvement/préparation d'échantillons de diagnostic clinique Prélèvement/préparation d'échantillons de sang
S-Monovette® is a blood collection system that combines the aspiration and the vacuum techniques.
  • Aspiration technique for routine blood collection
  • Vacuum technique is always available
  • For all ages, from young to old
Suitable for all vein conditions and achieves a sample quality for high quality results.