Imprimer
Erlenmeyer flasks, graduated, with screw cap
Fournisseur: Bellco Glass
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
BELC2515-02000EA 628 EUR
BELC2515-02000 BELC2515-04000
Erlenmeyer flasks, graduated, with screw cap
Ballons Flacons Erlenmeyers
These flasks are made of borosilicate type 1, class A with 45 mm PP screw caps and pouring rings.
  • Provides flexible, secure seal without a liner
  • Graduated to approximate volumes
  • Ideal for all cell culture and media storage purposes