Imprimer
Tubing connectors, straight, HDPE
Fournisseur: Semadeni
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
229-7486EA 5.4 EUR
229-7486
Tubing connectors, straight, HDPE
Connecteurs Raccords pour tuyaux
Tubing connector, made of HDPE, with taper ends.
  • Good temperature resistance
  • Good chemical resistance