Imprimer
Tubing connectors, Y-shaped, TPX
Fournisseur: Semadeni
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
229-4621EA 33.3 EUR
229-4621 229-4623 229-4622
Tubing connectors, Y-shaped, TPX
Raccords et connecteurs
Tubing connectors, Y-shaped, made of TPX, transparent.
  • Strong design with long, conical connections
  • Temperature resistant up to 130 °C
  • Sterilisable