Imprimer
TLC plates, cellulose MN 300
Fournisseur: MACHEREY-NAGEL
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
POLYGRAM® ALUGRAM®
552-0313EA 141 EUR
552-0313 552-0309 552-0132 552-0130 552-0174 552-0173 552-0312 552-0311 552-0179 552-0177 552-0133 552-0183 552-0182 552-0181 552-1290
TLC plates, cellulose MN 300
Plaques CCM
These native fibrous cellulose layers are available as glass plates, POLYGRAM® polyester sheets and ALUGRAM® aluminium sheets. They are used in partition chromatography of polar substances such as for the separations of amino acids, carboxylic acids or carbohydrates.
  • Alternative selectivity to silica 60
Celluolse MN 300: Fibre length (95%) 2 to 20 μmg, average degree of polymerisation 400 to 500, specific surface acc. to Blaine 15000 cm²/g.