Imprimer
Dishes, evaporating, flat bottom, half-round
Fournisseur: Bohemia Cristal (Kavalierglass)
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
KAVA641322274501EA 15 EUR
KAVA641322274501 KAVA641322274801 KAVA641322274401 KAVA641322274301 KAVA641322274201 KAVA641322274991 KAVA641322274901 KAVA641322274701 KAVA641322274601 KAVA641322274101 KAVA641322274121
Dishes, evaporating, flat bottom, half-round
Plateaux/Récipients/Pots Capsules d'évaporation
Made of porcelain.
  • Very good chemical resistance
  • Heat resistant up to 1000 °C
  • Low thermal expansion
For a wide range of laboratory applications. With spout and flat bottom. White glaze inside and outside.