Imprimer
Thistle funnels, PP, Azlon®
Fournisseur: SciLabware
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
221-0046EA 140 EUR
221-0046
Thistle funnels, PP, Azlon®
Entonnoirs Entonnoirs pour la filtration Entonnoirs filtrants
Thistle shaped funnel, made of PP, with detachable long stem.
  • Stem can be cut to size
  • Good chemical resistance
  • Good temperature resistance