Imprimer

Buffer, sodium citrate, 0,5M solution, pH 5,5

Fournisseur: Thermo Fisher Scientific
Ratings: Ecrire un avis

Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5

J63199.APEA 111 EUR
J63199.AP J63199.AK
Buffer, sodium citrate, 0,5M solution, pH 5,5
Tampons Tampons biologiques
Storage temperature: 2 to 8 °C
Physical form: liquid
Appearance: colourless
Odour: odourless
MDL No.: MFCD00012462
Commander maintenant

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR