Imprimer
Buffers, Tris-HCl, 1M solutions, Ultrapure
Fournisseur: Affymetrix
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
USBM22638100MLEA 23.2 EUR
USBM22638100ML
Buffers, Tris-HCl, 1M solutions, Ultrapure
1M solution of Tris, Ultrapure in high purity deionised water. Solution is 0,2 μm filtered and dispensed into durable, square Nalgene bottles.
  • DNase (endo): Passes test
  • DNase (exo): Passes test
  • RNase: Passes test
  • Protease: Passes test