Imprimer

4,5-Dicyano-2-(2-pyridyl)imidazole, 97%

Ratings: Ecrire un avis

Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5

H54586.MDEA 33.5 EUR
ALFAH54586.MD ALFAH54586.06 ALFAH54586.03
4,5-Dicyano-2-(2-pyridyl)imidazole, 97%
Purity: 97%
CAS No.: 58259-78-2
Molecular Formula: C10H5N5
Formula Weight: 195.18
Beilstein Reference: 17971891
UN No.: 3439
MDL No.: MFCD20265297
Commander maintenant

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR