Imprimer

2-(tert-Butylamino)-4'-chloropropiophenone hydrochloride

Marques : U.S. Pharmacopeia
Ratings: Ecrire un avis

Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5

USPH1078744EA 1240 EUR
USPH1078744
2-(tert-Butylamino)-4'-chloropropiophenone hydrochloride
2-(tert-Butylamino)-4'-chloropropiophenone hydrochloride
Formule: C₁₃H₁₉Cl₂NO
Numéro CAS: 1049718-72-0

Commander maintenant

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR