Imprimer
Funnel holders, SP Bel-Art
Fournisseur: Bel-Art Products, a part of SP
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
BELAF182420000EA 88.5 EUR
BELAF182420000
Funnel holders, SP Bel-Art
Supports de statif
Non-corrosive polypropylene plastic funnel holder and clamp supports for funnels.
  • For funnels 25 to 152 mm Ø
  • With two bushings for 9,5 or12,7 mm Ø support rods
  • Removable inner ring for smaller funnels