Imprimer
PCR tubes, 0,2/0,5 ml, with flat cap
Fournisseur: Nolato Treff AG
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
96.10385.9.01 96.9852.5.01 96.9852.6.01
TREF96.10385.9.01EA 73.1 EUR
TREF96.10385.9.01 TREF96.9852.5.01 TREF96.9852.6.01
PCR tubes, 0,2/0,5 ml, with flat cap
Tubes Tubes PCR
Made of virgin PP resin.
  • High chemical resistance
  • Very good temperature resistance
  • Free of RNase, DNase, human DNA, PCR inhibitors, endotoxins and ATP