Imprimer
Multirisks secure cabinets
Fournisseur: Trionyx
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
TRIOASVM22EA 1910 EUR
TRIOASVM22
Multirisks secure cabinets
Mobilier Armoires de stockage
Multi risk secure cabinet insure to separate the dangerous product families.
  • White epoxy paint RAL 9010
  • Steel containment shelves adjustable in height
  • Built in steel 10/10th
  • One removable base trough in steel in the bottom of each compartment
  • Lockable doors
High and low vents, outlet with flame arresters and Ø 100 mm outlet for a possible external connection. Handling doors with key locking system.
Certifications: Large warning labels in compliance with ISO 3864 norm and the European directive 92/58/CEE.
Informations de livraison: Supplied with PVC equipment for 'Acids' compartment including PVC protection shelf and PVC retention tank, additional steel retention shelf, industrial absorbent mat, 'Toxic', 'Flammable', 'Acid' large warning label.