Imprimer
Disinfectant wipes, Bacillol® AF Tissues
Fournisseur: Bode
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
Bacillol® AF
115-2157EA 19 EUR
115-2157
Disinfectant wipes, Bacillol® AF Tissues
Lingettes Lingettes et essuie-mains à usage unique Lingettes désinfectantes
Bacillol® AF Tissues are pre-soaked, ready to use disinfectant wipes made from high quality, tear resistant and absorbent PET fleece. The aldehyde-, colourant- and fragrance-free wipes act rapidly and comprehensively. They are suitable for disinfection of alcohol resistant surfaces, such as work surfaces in medical practices, labs and clinics, and also medical equipment.
  • Can also be used without gloves thanks to the skin friendly formulation
  • Safe and easy dispensing of individual wipes from resealable flowpack
  • Broad range of applications and material compatibility
  • Bacillol® AF saturation fluid guarantees effective moistening
Microbiological effectiveness: bactericidal, yeasticidal, fungicidal, tuberculocidal, mycobactericidal, limited virucidal action (incl. HBV, HIV, HCV), MNV, adeno-, polyoma- and rotaviruses.
VAH, Robert Koch Institute (RKI) list of tested and recognised disinfectants and methods.
Cdt: 80 wipes per resealable, handy flowpack. A wall mounting for the pack is also available.
Attention: *Le(s) produit(s) sur cette page sont hors du champ d'application de la directive européenne 2019/904