Imprimer
High visibility jacket, Windstopper® 744 GWG
Fournisseur: Fristads Kansas Group
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
Windstopper®
KWIN100369271XSEA 0 EUR
KWIN100369271XS KWIN100369271L KWIN1003692712XL KWIN1003692713XL KWIN100369271M KWIN100369171M KWIN100369171XL KWIN1003691713XL KWIN1003691712XL KWIN100369171S KWIN100369271S KWIN100369171XS KWIN100369271XL
High visibility jacket, Windstopper® 744 GWG
Tenue de travail
Water repellent, windproof jacket made of 3-layer Windstopper® laminate (100% polyester). With a reflective detail on shoulders.
  • Front placket opening with zip to top of collar and snap fastening
  • Two front pockets with zip, one with D ring for plastic ID-card holder
  • Chest pocket with zip
  • Two inside pockets, one with loop for earpiece
  • Adjustable cuffs, pre-shaped elbows
  • Drawstring hem
  • ePTFE membrane
Weight: 265 g/m²
Certifications: EN 20471 (class 3)
OEKO-TEX® certified