Imprimer
Oak jacket, chainsaw protection, CH10
Fournisseur: Portwest
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
PRTWCH10BKRXXLEA 91.8 EUR
PRTWCH10BKRXXL PRTWCH10BKRXL PRTWCH10BKRS PRTWCH10BKRL
Oak jacket, chainsaw protection, CH10
Tenue de travail
Durable jacket made of 65% polyester and 35% cotton, with water-repellent finish. The clothing has been designed for extreme outdoor conditions and incorporates stretch fabrics in key areas for greater ease of movement.
  • Longer sleeves, full length zipper with storm flap
  • Cuffs with Velcro strap adjustments for greater freedom of movement
  • Several pockets
  • Perfect for professional chainsaw users
Weight: 245 g/m²