Imprimer
Flame-retardant bomber jacket, Waverly 7361
Fournisseur: Sioen
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
Sio-Start® FR
SIOE7361/B75/3XLEA 230 EUR
SIOE7361/B75/3XL SIOE7361/B75/L SIOE7361/B75/2XL SIOE7361/B75/M SIOE7361/B75/1XL SIOE7361/B75/S
Flame-retardant bomber jacket, Waverly 7361
Tenue de travail
Anti-static jacket made of 100% polyester fabric with 100% FR PU coating + AST. With fixed flame-retardant cotton lining and detachable flame-retardant fleece lining in body (without sleeves, can not be worn separately).
  • Straight collar with foldaway hood in
  • Zip closure under flap with touch & close fastening
  • Two patched pockets with flap, one Napoleon pocket, one inside pocket
  • Sleeve narrowing by press studs
  • Elasticated hem
  • Taped seams
  • Flame retardant reflective tape (25 mm)
The fabric is water repellent on the outside and features a moisture attracting coating on the inside.
Back length: 75 cm
Weight: 250 g/m²
Certifications: EN 13034 (Type PB/6), EN 531 (A/B2/C1), EN 14116 (inside index 3; outside index 1), EN 1149-5, EN 343 (3-3)
Detachable lining only: EN 14058 (1XXXX), EN ISO 14116 (Index 1)