Imprimer
AccuStart™ II Taq DNA polymerase
Fournisseur: Quantabio
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
AccuStart™
733-2256EA 2170 EUR
733-2256 733-2255 733-2258
AccuStart™ II Taq DNA polymerase
Réactifs pour acides nucléiques End-point PCR Enzymes and Kits
AccuStart™ II Taq DNA Polymerase is a high purity, recombinant Taq DNA polymerase preparation with high avidity monoclonal antibodies that bind the polymerase and keep it inactive prior to the initial PCR denaturation step. Upon heat activation (1 minute at 94 °C), the antibodies denature irreversibly, releasing fully active, unmodified Taq DNA polymerase. This enables specific and efficient primer extension with the convenience of room temperature reaction assembly. Non-specific extension of primers at low temperatures is a common cause of artifacts and poor sensitivity in PCR. The AccuStart™ II automatic hot-start enables specific and efficient primer extension in the PCR process with the added convenience of room temperature reaction assembly. The included 10X PCR Buffer II is a new optimised buffer that provides higher product yield, improved specificity, and enhanced multiplexing capability.
Activated AccuStart™ II Taq DNA Polymerase possesses 5'➝3' DNA polymerase activity and a double-strand specific 5'➝3' exonuclease. The polymerase does not have 3'-exonuclease activity and is free of any contaminating endo or exonuclease activities. One unit is defined as the amount of enzyme that will incorporate 10 nmol of dNTP into acid-insoluble material in 30 minutes at 74 °C. AccuStart™ II Taq DNA Polymerase contains extremely low levels of residual host, E. coli genomic DNA.